Assay Details
| Gene Symbol: BTG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006806.4 | NP_006797.3 | 3 - 4 | 570 | 88 | |
| XM_017028265.1 | XP_016883754.1 | 4 - 5 | 708 | 88 | ||
| XM_017028266.1 | XP_016883755.1 | 4 - 5 | 665 | 88 | ||
| GenBank mRNA | AK289408.1 | - | 3 - 4 | 518 | 88 | |
| CR457061.1 | - | 2 - 3 | 310 | 88 | ||
| D64110.1 | - | 3 - 4 | 404 | 88 | ||
| DR155504.1 | - | 3 - 4 | 486 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000299.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003588.hg.1 | ENST00000339775.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003588.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003588.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003588.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003588.hg.1 | TC21000299.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | TC2100007663.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151953.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |

BTG3
Genomic Map