Assay Details
| Gene Symbol: ABCC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF418600.1 | - | 7 - 8 | 1129 | 109 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000429114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000429114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000429114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000429114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000429114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000429114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000429114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000429114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000429114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000429114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000463099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000467409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000471902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000481582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | ENST00000482980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006586.hg.1 | NR_003088.1 | Refseq |

ABCC13
Genomic Map