Assay Details
Gene Symbol: NT5C3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166118.2 | NP_001159590.1 | 3 - 4 | 436 | 148 | |
XM_011515409.2 | XP_011513711.1 | 3 - 4 | 829 | 148 | ||
GenBank mRNA | CB127877.1 | - | 3 - 4 | 175 | 148 | |
DB243020.1 | - | 3 - 4 | 180 | 148 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001264.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001166118.2,XM_011515409.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000449201.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000449201.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000449201.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000449201.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000449201.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000473083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000473083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000473083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000497542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000497542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000497542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000497542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
