Assay Details
Gene Symbol: NT5C3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002009.2 | NP_001002009.1 | 6 - 7 | 586 | 139 | |
NM_001002010.2 | NP_001002010.1 | 5 - 6 | 531 | 139 | ||
NM_001166118.2 | NP_001159590.1 | 7 - 8 | 736 | 139 | ||
NM_016489.12 | NP_057573.2 | 6 - 7 | 650 | 139 | ||
XM_011515409.2 | XP_011513711.1 | 7 - 8 | 1129 | 139 | ||
GenBank mRNA | AF151067.1 | - | 5 - 6 | 372 | 139 | |
AF312735.1 | - | 5 - 6 | 467 | 139 | ||
AK290118.1 | - | 5 - 6 | 488 | 139 | ||
AK314109.1 | - | 5 - 6 | 530 | 139 | ||
AL136716.1 | - | 5 - 6 | 496 | 139 | ||
BC013292.2 | - | 5 - 6 | 414 | 139 | ||
BC015856.1 | - | 6 - 7 | 395 | 4669009 | 139 | |
BC066914.1 | - | 6 - 7 | 511 | 5264738 | 139 | |
BC071652.1 | - | 5 - 6 | 513 | 3892369 | 139 | |
CB127877.1 | - | 7 - 8 | 475 | 139 | ||
CR533518.1 | - | 5 - 6 | 454 | 139 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001264.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000242210.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000381626.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000396152.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000405342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409467.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000409787.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000449201.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000449201.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000449201.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000449201.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000449201.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000456458.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000461851.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000464840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000473083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000473083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000473083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000497542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000497542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000497542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000497542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000610140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | ENST00000620705.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002009.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001002010.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_001166118.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010692.hg.1 | NM_016489.12 | Refseq |
