Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018416.hg.1 |
TC02001198.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018416.hg.1 |
ENST00000261016.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018416.hg.1 |
ENST00000261017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018416.hg.1 |
ENST00000261018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018416.hg.1 |
ENST00000295851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018416.hg.1 |
ENST00000422511.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018416.hg.1 |
ENST00000424558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018416.hg.1 |
ENST00000430418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018416.hg.1 |
ENST00000430574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018416.hg.1 |
NM_005759.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000261016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000261017.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000261018.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000295851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000411547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000417864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000422511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000422719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000424558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000430418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000430574.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000441061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000451591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000454023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
NM_001282925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
NM_001282926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
NM_001282927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
NM_001282932.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
NM_005759.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000261016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000261017.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000261018.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000295851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000411547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000417864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000422511.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000422719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000424558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000430418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000430574.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000441061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000451591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
ENST00000454023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
NM_001282925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
NM_001282926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
NM_001282927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
NM_001282932.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
NM_005759.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174525.hg.1 |
TC0200010518.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000261018.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000295851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000411547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000417864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000424558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000430418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
NM_001282925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
NM_001282926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000261018.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000295851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000411547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000417864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000424558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
ENST00000430418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
NM_001282925.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
NM_001282926.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174527.hg.1 |
TC0200010518.hg.1 |
Clariom D TC |