Assay Details
| Gene Symbol: TRAF3IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015650.3 | NP_056465.2 | 9 - 10 | 1377 | 61 | |
| XM_011510944.2 | XP_011509246.1 | 10 - 11 | 1534 | 61 | ||
| XM_011510945.2 | XP_011509247.1 | 10 - 11 | 1495 | 61 | ||
| XM_011510946.2 | XP_011509248.1 | 9 - 10 | 1462 | 61 | ||
| XM_011510947.2 | XP_011509249.1 | 10 - 11 | 1521 | 61 | ||
| XM_011510948.2 | XP_011509250.1 | 8 - 9 | 1336 | 61 | ||
| XM_011510950.2 | XP_011509252.1 | 2 - 3 | 337 | 61 | ||
| GenBank mRNA | AF242456.1 | - | 9 - 10 | 1476 | 61 | |
More Information
Set Membership: |
|

TRAF3IP1
Genomic Map