Assay Details
| Gene Symbol: LRP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005267510.1 | XP_005267567.1 | 4 - 5 | 1101 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRP10| Gene Symbol: LRP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005267510.1 | XP_005267567.1 | 4 - 5 | 1101 | 56 | |
Set Membership: |
|