Assay Details
| Gene Symbol: NDRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282215.1 | NP_001269144.1 | 12 - 13 | 880 | 107 | |
| GenBank mRNA | BC011240.1 | - | 12 - 13 | 840 | 4153602 | 107 |
More Information
Set Membership: |
|

NDRG2
Genomic Map