Assay Details
| Gene Symbol: MAP1LC3B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011538652.2 | XP_011536954.1 | 1 | 54 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 173 | LINC00173 | NCRNA00173 | -22.4kb | Anti-sense | Chr. 12 - 116533422 - 116536513 on NCBI Build GRCh38 |
| Downstream | chromosome 12 open reading frame 49 | C12orf49 | 153.8kb | Sense | Chr. 12 - 116713320 - 116738070 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011538652.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MAP1LC3B2
Genomic Map