Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010223.hg.1 |
ENST00000356173.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010223.hg.1 |
ENST00000375523.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010223.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010223.hg.1 |
TC20000767.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161801.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161801.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161801.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161801.hg.1 |
TC2000008870.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000161801.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161801.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161801.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000356173.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375519.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000375523.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
ENST00000420875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008870.hg.1 |
NM_080675.3 |
Refseq |