Assay Details
| Gene Symbol: RCBTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268.3 | NP_001259.1 | 2 - 3 | 331 | 84 | |
| NM_001286830.1 | NP_001273759.1 | 2 - 3 | 331 | 84 | ||
| XM_017020369.1 | XP_016875858.1 | 2 - 3 | 280 | 84 | ||
| XM_017020371.1 | XP_016875860.1 | 2 - 3 | 415 | 84 | ||
| XM_017020374.1 | XP_016875863.1 | 2 - 3 | 280 | 84 | ||
| XM_017020375.1 | XP_016875864.1 | 2 - 3 | 280 | 84 | ||
| XM_017020376.1 | XP_016875865.1 | 2 - 3 | 280 | 84 | ||
| XM_017020377.1 | XP_016875866.1 | 2 - 3 | 280 | 84 | ||
| XM_017020378.1 | XP_016875867.1 | 2 - 3 | 280 | 84 | ||
| XM_017020379.1 | XP_016875868.1 | 2 - 3 | 280 | 84 | ||
| GenBank mRNA | AF060219.1 | - | 1 - 2 | 112 | 84 | |
| AK023010.1 | - | 1 - 2 | 74 | 84 | ||
| AK301516.1 | - | 2 - 3 | 155 | 84 | ||
| AK315703.1 | - | 2 - 3 | 251 | 84 | ||
| BC029052.1 | - | 2 - 3 | 135 | 5194795 | 84 | |
| DB497652.1 | - | 2 - 3 | 330 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF060219.1,AK023010.1,AK301516.1,AK315703.1,BC029052.1,NM_001268.3,NM_001286830.1,XM_017020369.1,XM_017020371.1,XM_017020374.1,XM_017020375.1,XM_017020376.1,XM_017020377.1,XM_017020378.1,XM_017020379.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

RCBTB2
Genomic Map