Assay Details
| Gene Symbol: UTS2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011512631.1 | XP_011510933.1 | 1 - 2 | 197 | 101 | |
| XM_017006091.1 | XP_016861580.1 | 1 - 2 | 197 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011512631.1, XM_017006091.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000464814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000464814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000464814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000490825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000490825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000490825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000490825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | ENST00000490825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000340524.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000425357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000427544.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000432514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000440476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000446788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000463450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000464814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000464814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000464814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000490825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000490825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000490825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000490825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | ENST00000490825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013532.hg.1 | NM_198152.3 | Refseq |

UTS2B
Genomic Map