Assay Details
| Gene Symbol: SKIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145097.2 | NP_001138569.1 | 3 - 4 | 1997 | 92 | |
| GenBank mRNA | U70730.1 | - | 3 - 4 | 1997 | 92 | |
More Information
Set Membership: |
|

SKIL
Genomic Map