Assay Details
| Gene Symbol: POPDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK377176.1 | - | 3 - 4 | 341 | 81 | |
| AK377180.1 | - | 3 - 4 | 341 | 81 | ||
| AK377181.1 | - | 3 - 4 | 341 | 81 | ||
| AK377183.1 | - | 3 - 4 | 341 | 81 | ||
| AK377184.1 | - | 3 - 4 | 341 | 81 | ||
More Information
Set Membership: |
|

POPDC2
Genomic Map