Assay Details
Gene Symbol: AK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000476.2 | NP_000467.1 | 1 - 2 | 123 | 146 | |
GenBank mRNA | DA077967.1 | - | 1 - 2 | 123 | 147 | |
EU794628.1 | - | 1 - 2 | 123 | 146 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09002920.hg.1
Additional Information:
This assay is in the 5'UTR region of the EU794628.1,NM_000476.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nucleoside diphosphate phosphorylation
ADP biosynthetic process
cell cycle arrest
nucleoside triphosphate biosynthetic process
nucleobase-containing small molecule interconversion
AMP metabolic process
ATP metabolic process
adenylate kinase activity
nucleoside diphosphate kinase activity
ATP binding
nucleoside diphosphate phosphorylation
ADP biosynthetic process
cell cycle arrest
nucleoside triphosphate biosynthetic process
nucleobase-containing small molecule interconversion
AMP metabolic process
ATP metabolic process
adenylate kinase activity
nucleoside diphosphate kinase activity
ATP binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09020297.hg.1 | TC09002920.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09020297.hg.1 | ENST00000373176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020297.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09020294.hg.1 | TC09002920.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09020294.hg.1 | ENST00000373156.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020294.hg.1 | ENST00000373176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020294.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09020296.hg.1 | TC09002920.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09020296.hg.1 | ENST00000373176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020296.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09020298.hg.1 | TC09002920.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09020298.hg.1 | ENST00000373176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020298.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186722.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186722.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186722.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186722.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186722.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186722.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186722.hg.1 | TC0900012277.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900186725.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186725.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186725.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186725.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186725.hg.1 | TC0900012277.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000223836.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000413016.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012277.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186720.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186720.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186720.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186720.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186720.hg.1 | ENST00000373156.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186720.hg.1 | ENST00000373176.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186720.hg.1 | ENST00000550992.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186720.hg.1 | NM_000476.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186720.hg.1 | TC0900012277.hg.1 | Clariom D TC |
