Assay Details
| Gene Symbol: GNAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002069.5 | NP_002060.4 | 1 - 2 | 456 | 107 | |
| GenBank mRNA | AF055013.1 | - | 1 - 2 | 131 | 107 | |
| AF493905.1 | - | 1 - 2 | 119 | 107 | ||
| AK292953.1 | - | 1 - 2 | 430 | 107 | ||
| AL049933.1 | - | 1 - 2 | 434 | 107 | ||
| BC026326.1 | - | 1 - 2 | 455 | 4791696 | 107 | |
| M17219.1 | - | 1 - 2 | 104 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000508.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
cell cycle
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
response to peptide hormone
regulation of cAMP-mediated signaling
negative regulation of synaptic transmission
cell division
cellular response to forskolin
magnesium ion binding
G-protein coupled receptor binding
GTPase activity
signal transducer activity
protein binding
GTP binding
GDP binding
G-protein beta/gamma-subunit complex binding
G-protein coupled serotonin receptor binding
GTPase activating protein binding
protein folding
cell cycle
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
response to peptide hormone
regulation of cAMP-mediated signaling
negative regulation of synaptic transmission
cell division
cellular response to forskolin
magnesium ion binding
G-protein coupled receptor binding
GTPase activity
signal transducer activity
protein binding
GTP binding
GDP binding
G-protein beta/gamma-subunit complex binding
G-protein coupled serotonin receptor binding
GTPase activating protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000418742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000418742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000418742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000418742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000418742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000447650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000447650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000447650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007178.hg.1 | TC07000508.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007178.hg.1 | ENST00000351004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007178.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007178.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007179.hg.1 | TC07000508.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007179.hg.1 | ENST00000351004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007179.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007179.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007176.hg.1 | TC07000508.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007176.hg.1 | ENST00000351004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007176.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156859.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156859.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156859.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156859.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156859.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156859.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156859.hg.1 | TC0700008181.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000418742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000418742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000418742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000418742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000418742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000447650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000447650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000447650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000457358.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_001256414.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008181.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156857.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156857.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156857.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156857.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156857.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156857.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156857.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156857.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156857.hg.1 | TC0700008181.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700156858.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156858.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156858.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156858.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156858.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156858.hg.1 | ENST00000442586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156858.hg.1 | ENST00000490206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156858.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156858.hg.1 | TC0700008181.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700156855.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156855.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156855.hg.1 | ENST00000351004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156855.hg.1 | NM_002069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156855.hg.1 | TC0700008181.hg.1 | Clariom D TC |

GNAI1
Genomic Map