Assay Details
Gene Symbol: TXK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003328.2 | NP_003319.2 | 2 - 3 | 159 | 61 | |
GenBank mRNA | DB103575.1 | - | 2 - 3 | 153 | 61 | |
L27071.1 | - | 2 - 3 | 159 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001173.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
adaptive immune response
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
activation of phospholipase C activity
integrin-mediated signaling pathway
regulation of platelet activation
cell differentiation
positive regulation of interferon-gamma production
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
tissue regeneration
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
T cell receptor signaling pathway
positive regulation of interferon-gamma-mediated signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II regulatory region DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
cytokine production
adaptive immune response
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
activation of phospholipase C activity
integrin-mediated signaling pathway
regulation of platelet activation
cell differentiation
positive regulation of interferon-gamma production
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
tissue regeneration
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
T cell receptor signaling pathway
positive regulation of interferon-gamma-mediated signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II regulatory region DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04016145.hg.1 | TC04001173.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04016145.hg.1 | ENST00000264316.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016145.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016145.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000507351.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000509681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | ENST00000514937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010615.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172438.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172438.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172438.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172438.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172438.hg.1 | ENST00000264316.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172438.hg.1 | ENST00000506073.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172438.hg.1 | ENST00000510457.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172438.hg.1 | NM_003328.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172438.hg.1 | TC0400010615.hg.1 | Clariom D TC |
