Assay Details
| Gene Symbol: UGT2B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005265702.3 | XP_005265759.1 | 2 - 3 | 471 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005265702.3 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |

UGT2B7
Genomic Map