Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099286.2 | NP_001092756.1 | 1 - 2 | 364 | 148 | |
| XM_011535410.2 | XP_011533712.1 | 1 - 2 | 328 | 148 | ||
| GenBank mRNA | AK292303.1 | - | 1 - 2 | 315 | 148 | |
| DB016393.1 | - | 1 - 2 | 340 | 148 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002142.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK292303.1,NM_001099286.2,XM_011535410.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06026961.hg.1 | TC06002142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026961.hg.1 | ENST00000420702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026961.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026961.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026960.hg.1 | TC06002142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026960.hg.1 | ENST00000420702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026960.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026960.hg.1 | ENST00000451457.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026960.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026960.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191791.hg.1 | TC0600013271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600191792.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191792.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191792.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191792.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191792.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191792.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191792.hg.1 | TC0600013271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |

MTFR2
Genomic Map