Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099286.2 | NP_001092756.1 | 3 - 4 | 585 | 125 | |
| NM_001318738.1 | NP_001305667.1 | 4 - 5 | 397 | 125 | ||
| NM_138419.4 | NP_612428.2 | 3 - 4 | 297 | 125 | ||
| XM_011535410.2 | XP_011533712.1 | 4 - 5 | 649 | 125 | ||
| XM_011535412.2 | XP_011533714.1 | 2 - 3 | 390 | 125 | ||
| XM_011535413.2 | XP_011533715.1 | 2 - 3 | 3504 | 125 | ||
| GenBank mRNA | AK125758.1 | - | 4 - 5 | 354 | 125 | |
| AK292303.1 | - | 3 - 4 | 536 | 125 | ||
| BC063688.1 | - | 3 - 4 | 250 | 4455541 | 125 | |
| BG831782.1 | - | 3 - 4 | 288 | 125 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002142.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026955.hg.1 | TC06002142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026955.hg.1 | ENST00000420702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026955.hg.1 | ENST00000451457.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026955.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026955.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026954.hg.1 | TC06002142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026954.hg.1 | ENST00000420702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026954.hg.1 | ENST00000451457.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026954.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026954.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191785.hg.1 | TC0600013271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |

MTFR2
Genomic Map