Assay Details
| Gene Symbol: LINC00571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_047500.1 | 4 - 5 | 396 | 111 | ||
| GenBank mRNA | BM023585.1 | - | 4 - 5 | 407 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000454060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000454060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000454060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | ENST00000454060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000454060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000454060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000454060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | ENST00000454060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008651.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158448.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158448.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158448.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158448.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158448.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158448.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158448.hg.1 | TC1300008651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300158459.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158459.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158459.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158459.hg.1 | ENST00000454060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158459.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158459.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158459.hg.1 | ENST00000434565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158459.hg.1 | ENST00000451826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158459.hg.1 | ENST00000454060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158459.hg.1 | NR_047500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158459.hg.1 | TC1300008651.hg.1 | Clariom D TC |

LINC00571
Genomic Map