Assay Details
| Gene Symbol: LANCL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110604.1 | 5 - 6 | 516 | 83 | ||
| NR_110605.1 | 5 - 6 | 489 | 83 | |||
| NR_110606.1 | 4 - 5 | 331 | 83 | |||
| GenBank mRNA | AL599033.1 | - | 5 - 6 | 459 | 83 | |
| DA820426.1 | - | 4 - 5 | 331 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175612.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175612.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175612.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175612.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175612.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175612.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175612.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175612.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175612.hg.1 | TC0200010655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200175614.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175614.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175614.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175614.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175614.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175614.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175614.hg.1 | TC0200010655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000420418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | ENST00000433296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010655.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175611.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175611.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175611.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175611.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175611.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175611.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175611.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175611.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175611.hg.1 | TC0200010655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200175613.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175613.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175613.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175613.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175613.hg.1 | ENST00000416344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175613.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175613.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175613.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175613.hg.1 | TC0200010655.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200175615.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175615.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175615.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175615.hg.1 | NR_110604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175615.hg.1 | NR_110605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175615.hg.1 | NR_110606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175615.hg.1 | TC0200010655.hg.1 | Clariom D TC |

LANCL1-AS1
Genomic Map