Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318738.1 | NP_001305667.1 | 1 - 2 | 74 | 135 | |
| NM_138419.4 | NP_612428.2 | 1 - 2 | 74 | 135 | ||
| GenBank mRNA | BG831782.1 | - | 1 - 2 | 65 | 135 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002142.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001318738.1,NM_138419.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026967.hg.1 | TC06002142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026967.hg.1 | ENST00000420702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026967.hg.1 | ENST00000451457.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026967.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026967.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026959.hg.1 | TC06002142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026959.hg.1 | ENST00000420702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026959.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026959.hg.1 | ENST00000451457.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026959.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026959.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012793.hg.1 | ENST00000438685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191793.hg.1 | TC0600013271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012793.hg.1 | ENST00000438685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191790.hg.1 | TC0600013271.hg.1 | Clariom D TC |

MTFR2
Genomic Map