Assay Details
Gene Symbol: OSMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001323505.1 | NP_001310434.1 | 13 - 14 | 2154 | 149 | |
NM_001323506.1 | NP_001310435.1 | 13 - 14 | 2157 | 149 | ||
NM_003999.2 | NP_003990.1 | 13 - 14 | 2274 | 149 | ||
XM_005248384.1 | XP_005248441.1 | 13 - 14 | 2264 | 149 | ||
XM_011514161.2 | XP_011512463.1 | 13 - 14 | 2264 | 149 | ||
XM_017010019.1 | XP_016865508.1 | 13 - 14 | 2261 | 149 | ||
XM_017010020.1 | XP_016865509.1 | 7 - 8 | 964 | 149 | ||
GenBank mRNA | AK304016.1 | - | 8 - 9 | 1282 | 149 | |
BC125209.1 | - | 13 - 14 | 2044 | 40123501 | 149 | |
BC125210.1 | - | 13 - 14 | 2044 | 40123502 | 149 | |
U60805.1 | - | 13 - 14 | 2239 | 149 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000179.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05002111.hg.1 | TC05000179.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05002111.hg.1 | ENST00000274276.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05002111.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05002110.hg.1 | TC05000179.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05002110.hg.1 | ENST00000274276.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05002110.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148738.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148738.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148738.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148738.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148738.hg.1 | TC0500007201.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500148739.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148739.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148739.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500148739.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500148739.hg.1 | TC0500007201.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
