Assay Details
Gene Symbol: MITD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320417.1 | NP_001307346.1 | 3 - 4 | 597 | 151 | |
NM_001320418.1 | NP_001307347.1 | 3 - 4 | 473 | 151 | ||
NM_138798.2 | NP_620153.1 | 3 - 4 | 473 | 151 | ||
XM_011510581.2 | XP_011508883.1 | 3 - 4 | 485 | 151 | ||
XM_011510582.2 | XP_011508884.1 | 3 - 4 | 485 | 151 | ||
XM_011510583.1 | XP_011508885.1 | 3 - 4 | 571 | 151 | ||
XM_017003315.1 | XP_016858804.1 | 3 - 4 | 485 | 151 | ||
GenBank mRNA | BC018453.1 | - | 3 - 4 | 441 | 4509694 | 151 |
BU507044.1 | - | 3 - 4 | 455 | 151 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002132.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
cell separation after cytokinesis
negative regulation of protein binding
viral budding via host ESCRT complex
multivesicular body sorting pathway
protein binding
protein domain specific binding
phosphatidylinositol binding
protein homodimerization activity
mitotic cytokinesis
cell separation after cytokinesis
negative regulation of protein binding
viral budding via host ESCRT complex
multivesicular body sorting pathway
protein binding
protein domain specific binding
phosphatidylinositol binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02034715.hg.1 | TC02002132.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02034715.hg.1 | ENST00000289359.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02034715.hg.1 | ENST00000466880.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02034715.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000413710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000413710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000413710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000413710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000413710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000487588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000487588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000487588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | ENST00000487588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000409107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000413710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000413710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000413710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000413710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000413710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000438121.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000483721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000487588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000487588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000487588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | ENST00000487588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013603.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | ENST00000487588.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | ENST00000289359.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | ENST00000422537.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | ENST00000464685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | ENST00000466880.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | ENST00000487588.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | NM_138798.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200199801.hg.1 | TC0200013603.hg.1 | Clariom D TC |
