Assay Details
| Gene Symbol: PGM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018290.3 | NP_060760.2 | 5 - 6 | 603 | 125 | |
| XM_011513711.2 | XP_011512013.1 | 6 - 7 | 706 | 125 | ||
| GenBank mRNA | AF109360.1 | - | 5 - 6 | 543 | 125 | |
| AK001845.1 | - | 5 - 6 | 546 | 125 | ||
| AK223237.1 | - | 5 - 6 | 545 | 125 | ||
| AK297752.1 | - | 3 - 4 | 340 | 125 | ||
| AK303374.1 | - | 6 - 7 | 711 | 125 | ||
| AK310927.1 | - | 6 - 7 | 722 | 125 | ||
| AL136705.1 | - | 5 - 6 | 560 | 125 | ||
| BC010087.2 | - | 5 - 6 | 543 | 2964390 | 125 | |
| CR457274.1 | - | 5 - 6 | 530 | 125 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000213.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04003143.hg.1 | TC04000213.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003143.hg.1 | ENST00000381967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003143.hg.1 | ENST00000537241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003143.hg.1 | ENST00000544359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003143.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149486.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149486.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149486.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149486.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149486.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149486.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149486.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149486.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149486.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149486.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149486.hg.1 | TC0400007243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |

PGM2
Genomic Map