Assay Details
| Gene Symbol: PGM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018290.3 | NP_060760.2 | 8 - 9 | 1073 | 67 | |
| XM_011513711.2 | XP_011512013.1 | 9 - 10 | 1176 | 67 | ||
| GenBank mRNA | AF109360.1 | - | 8 - 9 | 1013 | 67 | |
| AK001845.1 | - | 8 - 9 | 1016 | 67 | ||
| AK223237.1 | - | 8 - 9 | 1015 | 67 | ||
| AK297752.1 | - | 6 - 7 | 810 | 67 | ||
| AK303374.1 | - | 9 - 10 | 1181 | 67 | ||
| AK310927.1 | - | 8 - 9 | 1002 | 67 | ||
| AL136705.1 | - | 8 - 9 | 1030 | 67 | ||
| BC010087.2 | - | 8 - 9 | 1013 | 2964390 | 67 | |
| CR457274.1 | - | 8 - 9 | 1000 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000213.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04003146.hg.1 | TC04000213.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003146.hg.1 | ENST00000381967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003146.hg.1 | ENST00000537241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003146.hg.1 | ENST00000544359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003146.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149489.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149489.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149489.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149489.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149489.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149489.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149489.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149489.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149489.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149489.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149489.hg.1 | TC0400007243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |

PGM2
Genomic Map