Assay Details
| Gene Symbol: PPP1R36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172365.2 | NP_758953.1 | 5 - 6 | 465 | 68 | |
| XM_005267354.3 | XP_005267411.1 | 5 - 6 | 385 | 68 | ||
| XM_005267355.3 | XP_005267412.1 | 4 - 5 | 312 | 68 | ||
| XM_011536468.2 | XP_011534770.1 | 5 - 6 | 433 | 68 | ||
| GenBank mRNA | AK058044.1 | - | 5 - 6 | 465 | 68 | |
| BC068989.1 | - | 5 - 6 | 430 | 30389944 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000392.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14004480.hg.1 | ENST00000298705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004480.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004480.hg.1 | TC14000392.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152013.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152013.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152013.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152013.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152013.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152013.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152013.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152013.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152013.hg.1 | TC1400007444.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |

PPP1R36
Genomic Map