Assay Details
Gene Symbol: PPP1R36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172365.2 | NP_758953.1 | 6 - 7 | 529 | 124 | |
XM_005267354.3 | XP_005267411.1 | 6 - 7 | 449 | 124 | ||
XM_005267355.3 | XP_005267412.1 | 5 - 6 | 376 | 124 | ||
XM_011536468.2 | XP_011534770.1 | 6 - 7 | 497 | 124 | ||
GenBank mRNA | AK058044.1 | - | 6 - 7 | 529 | 124 | |
BC068989.1 | - | 6 - 7 | 494 | 30389944 | 124 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000392.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR14004481.hg.1 | ENST00000298705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004481.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14004481.hg.1 | TC14000392.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152015.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152015.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152015.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152015.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152015.hg.1 | ENST00000298705.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152015.hg.1 | ENST00000467261.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152015.hg.1 | ENST00000557202.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152015.hg.1 | NM_172365.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152015.hg.1 | TC1400007444.hg.1 | Clariom D TC |
