Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000543743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000543743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006097.hg.1 |
ENST00000443185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006097.hg.1 |
ENST00000524431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006097.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006097.hg.1 |
TC18000581.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006102.hg.1 |
ENST00000443185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006102.hg.1 |
ENST00000524431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006102.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006102.hg.1 |
TC18000581.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006098.hg.1 |
ENST00000443185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006098.hg.1 |
ENST00000524431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006098.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006098.hg.1 |
TC18000581.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000613453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007721.hg.1 |
ENST00000613453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162239.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162239.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162239.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162239.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162239.hg.1 |
TC1800009086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800162239.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162239.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162239.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162239.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162238.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162238.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162238.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162238.hg.1 |
TC1800009086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800162238.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162238.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162238.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162240.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162240.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162240.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162240.hg.1 |
TC1800009086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800162240.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162240.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162240.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000581996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000613453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007721.hg.1 |
ENST00000613453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000443185.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000532857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000542818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000543743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000543743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
ENST00000617840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009086.hg.1 |
NM_014643.3 |
Refseq |