Assay Details
| Gene Symbol: SORBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF090937.1 | - | 1 | 1257 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PDZ and LIM domain 3 | PDLIM3 | ALP | -60.9kb | Sense | Chr. 4 - 185500660 - 185535606 on NCBI Build GRCh38 |
| Downstream | toll like receptor 3 | TLR3 |
CD283, IIAE2 |
471.0kb | Anti-sense | Chr. 4 - 186069155 - 186086724 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SORBS2
Genomic Map