Assay Details
| Gene Symbol: MBTPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015884.3 | NP_056968.1 | 6 - 7 | 987 | 95 | |
| GenBank mRNA | AF019612.1 | - | 6 - 7 | 889 | 95 | |
| AK292933.1 | - | 6 - 7 | 894 | 95 | ||
| AK307143.1 | - | 2 - 3 | 258 | 95 | ||
| AK312474.1 | - | 6 - 7 | 820 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000098.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cholesterol metabolic process
membrane protein intracellular domain proteolysis
response to endoplasmic reticulum stress
ATF6-mediated unfolded protein response
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
metalloendopeptidase activity
metallopeptidase activity
metal ion binding
cholesterol metabolic process
membrane protein intracellular domain proteolysis
response to endoplasmic reticulum stress
ATF6-mediated unfolded protein response
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
metalloendopeptidase activity
metallopeptidase activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000383912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_206923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_206923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_206923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_206923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_206923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006764.hg.1 | NM_206923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001325.hg.1 | TC0X000098.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001325.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001325.hg.1 | ENST00000379484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001325.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146325.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146325.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146325.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146325.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146325.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146325.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146325.hg.1 | TC0X00006764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000365779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000379484.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000383912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000429584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | ENST00000465888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_015884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_206923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_206923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_206923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_206923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_206923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006764.hg.1 | NM_206923.3 | Refseq |

MBTPS2
Genomic Map