Assay Details
| Gene Symbol: DDX21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256910.1 | NP_001243839.1 | 13 - 14 | 2263 | 90 | |
| NM_004728.3 | NP_004719.2 | 13 - 14 | 2144 | 90 | ||
| XM_011540336.1 | XP_011538638.1 | 12 - 13 | 1974 | 90 | ||
| XM_017016910.1 | XP_016872399.1 | 12 - 13 | 1892 | 90 | ||
| GenBank mRNA | AB062394.1 | - | 5 - 6 | 623 | 90 | |
| AK025629.1 | - | 10 - 11 | 1446 | 90 | ||
| AK315585.1 | - | 13 - 14 | 2093 | 90 | ||
| BC008071.2 | - | 13 - 14 | 2082 | 2987980 | 90 | |
| BX648405.1 | - | 13 - 14 | 2030 | 90 | ||
| CR749598.1 | - | 13 - 14 | 2264 | 90 | ||
| U41387.1 | - | 13 - 14 | 2095 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000413.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
rRNA processing
transcription from RNA polymerase II promoter
response to virus
RNA secondary structure unwinding
response to exogenous dsRNA
positive regulation of gene expression, epigenetic
double-stranded RNA binding
ATP-dependent RNA helicase activity
protein binding
ATP binding
rRNA binding
snoRNA binding
poly(A) RNA binding
7SK snRNA binding
osteoblast differentiation
rRNA processing
transcription from RNA polymerase II promoter
response to virus
RNA secondary structure unwinding
response to exogenous dsRNA
positive regulation of gene expression, epigenetic
double-stranded RNA binding
ATP-dependent RNA helicase activity
protein binding
ATP binding
rRNA binding
snoRNA binding
poly(A) RNA binding
7SK snRNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10004665.hg.1 | ENST00000354185.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004665.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004665.hg.1 | TC10000413.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007876.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153315.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153315.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153315.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153315.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153315.hg.1 | ENST00000354185.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153315.hg.1 | ENST00000620315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153315.hg.1 | NM_001256910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153315.hg.1 | NM_004728.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153315.hg.1 | TC1000007876.hg.1 | Clariom D TC |

DDX21
Genomic Map