| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000319363.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000459971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000459971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000459971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000459971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000477874.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000477874.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000477874.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000477874.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000477874.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000477874.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000477874.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
ENST00000612619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_001289905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006501.hg.1 |
NM_014339.6 |
Refseq |