| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16015842.hg.1 |
ENST00000219207.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16015842.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16015842.hg.1 |
TC16001137.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000564018.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000564018.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000564018.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000564018.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000564018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000564018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000564018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000564018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011546.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600193882.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600193882.hg.1 |
ENST00000564018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600193882.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600193882.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600193882.hg.1 |
ENST00000219207.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600193882.hg.1 |
ENST00000564018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600193882.hg.1 |
ENST00000569059.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600193882.hg.1 |
NM_015993.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600193882.hg.1 |
TC1600011546.hg.1 |
Clariom D TC |