Assay Details
| Gene Symbol: CTR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014633.4 | NP_055448.1 | 20 - 21 | 3006 | 89 | |
| GenBank mRNA | AK291794.1 | - | 20 - 21 | 2742 | 89 | |
| BC058914.1 | - | 20 - 21 | 2753 | 6527835 | 89 | |
| D63875.1 | - | 20 - 21 | 2666 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000187.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
endodermal cell fate commitment
inner cell mass cell differentiation
trophectodermal cell differentiation
blastocyst growth
transcription, DNA-templated
JAK-STAT cascade
histone monoubiquitination
Wnt signaling pathway
stem cell population maintenance
positive regulation of transcription elongation from RNA polymerase II promoter
histone H2B ubiquitination
negative regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
interleukin-6-mediated signaling pathway
cellular response to lipopolysaccharide
histone H3-K4 trimethylation
negative regulation of mRNA polyadenylation
regulation of genetic imprinting
positive regulation of histone H3-K79 methylation
positive regulation of histone H2B ubiquitination
RNA polymerase II core binding
protein binding
SH2 domain binding
negative regulation of transcription from RNA polymerase II promoter
endodermal cell fate commitment
inner cell mass cell differentiation
trophectodermal cell differentiation
blastocyst growth
transcription, DNA-templated
JAK-STAT cascade
histone monoubiquitination
Wnt signaling pathway
stem cell population maintenance
positive regulation of transcription elongation from RNA polymerase II promoter
histone H2B ubiquitination
negative regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
interleukin-6-mediated signaling pathway
cellular response to lipopolysaccharide
histone H3-K4 trimethylation
negative regulation of mRNA polyadenylation
regulation of genetic imprinting
positive regulation of histone H3-K79 methylation
positive regulation of histone H2B ubiquitination
RNA polymerase II core binding
protein binding
SH2 domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11002547.hg.1 | ENST00000361367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002547.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002547.hg.1 | TC11000187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000526874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000526874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000526874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000526874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000526874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000529355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000529355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000529355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000529355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000529898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000529898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000529898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | ENST00000529898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000524523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000526874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000526874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000526874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000526874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000526874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000529355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000529355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000529355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000529355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000529898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000529898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000529898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | ENST00000529898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006840.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148133.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148133.hg.1 | ENST00000529898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148133.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148133.hg.1 | ENST00000361367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148133.hg.1 | ENST00000529898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148133.hg.1 | NM_014633.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148133.hg.1 | TC1100006840.hg.1 | Clariom D TC |

CTR9
Genomic Map