Assay Details
| Gene Symbol: EPB41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166005.1 | NP_001159477.1 | 1 - 2 | 118 | 89 | |
| NM_203342.2 | NP_976217.1 | 1 - 2 | 118 | 89 | ||
| NM_203343.2 | NP_976218.1 | 1 - 2 | 118 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001166005.1, NM_203342.2, NM_203343.2 transcripts.
Set Membership: |
|

EPB41
Genomic Map