| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
ENST00000599339.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
ENST00000599339.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
ENST00000599339.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001344.hg.1 |
ENST00000221847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001344.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001344.hg.1 |
TC19000083.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900146837.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146837.hg.1 |
ENST00000599339.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146837.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146837.hg.1 |
TC1900006682.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900146837.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146837.hg.1 |
ENST00000599339.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146837.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
ENST00000221847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
ENST00000599339.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
ENST00000599339.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
ENST00000599339.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006682.hg.1 |
NM_005755.2 |
Refseq |