Assay Details
| Gene Symbol: PDE3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000922.3 | NP_000913.2 | 14 - 15 | 3236 | 98 | |
| XM_017017911.1 | XP_016873400.1 | 13 - 14 | 3154 | 98 | ||
| GenBank mRNA | AY459346.1 | - | 14 - 15 | 3174 | 98 | |
| BC136565.1 | - | 14 - 15 | 3129 | 9020555 | 98 | |
| BC136566.1 | - | 14 - 15 | 3129 | 9020556 | 98 | |
| BC144248.1 | - | 14 - 15 | 2976 | 9052768 | 98 | |
| BC150307.1 | - | 14 - 15 | 3236 | 8860429 | 98 | |
| X95520.1 | - | 14 - 15 | 2883 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000214.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cAMP catabolic process
negative regulation of cell adhesion
signal transduction
negative regulation of angiogenesis
cellular response to insulin stimulus
negative regulation of cell adhesion mediated by integrin
negative regulation of cAMP-mediated signaling
negative regulation of lipid catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
protein binding
protein kinase B binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
angiogenesis
cAMP catabolic process
negative regulation of cell adhesion
signal transduction
negative regulation of angiogenesis
cellular response to insulin stimulus
negative regulation of cell adhesion mediated by integrin
negative regulation of cAMP-mediated signaling
negative regulation of lipid catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
protein binding
protein kinase B binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11002875.hg.1 | ENST00000282096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002875.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002875.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002875.hg.1 | TC11000214.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000525439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000525439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000525439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000525439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000525439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000534317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000534317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | ENST00000534317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148725.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148725.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148725.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148725.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148725.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148725.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148725.hg.1 | TC1100006923.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000282096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000455098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000525439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000525439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000525439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000525439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000525439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000534317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000534317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | ENST00000534317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006923.hg.1 | NM_000922.3 | Refseq |

PDE3B
Genomic Map