Assay Details
| Gene Symbol: PDE3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000922.3 | NP_000913.2 | 2 - 3 | 1384 | 123 | |
| XM_006718249.3 | XP_006718312.1 | 2 - 3 | 1416 | 123 | ||
| XM_017017911.1 | XP_016873400.1 | 2 - 3 | 1416 | 123 | ||
| XM_017017912.1 | XP_016873401.1 | 2 - 3 | 1416 | 123 | ||
| GenBank mRNA | AY459346.1 | - | 2 - 3 | 1322 | 123 | |
| BC136565.1 | - | 2 - 3 | 1277 | 9020555 | 123 | |
| BC136566.1 | - | 2 - 3 | 1277 | 9020556 | 123 | |
| BC144248.1 | - | 2 - 3 | 1277 | 9052768 | 123 | |
| BC150307.1 | - | 2 - 3 | 1384 | 8860429 | 123 | |
| X95520.1 | - | 2 - 3 | 1031 | 123 | ||
More Information
Set Membership: |
|

PDE3B
Genomic Map