Assay Details
| Gene Symbol: COMTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144589.2 | NP_653190.2 | 4 - 5 | 530 | 150 | |
| GenBank mRNA | AK074421.1 | - | 4 - 5 | 483 | 150 | |
| AK290452.1 | - | 4 - 5 | 584 | 150 | ||
| AK302283.1 | - | 4 - 5 | 115 | |||
| AY358476.1 | - | 4 - 5 | 529 | 150 | ||
| BC023663.2 | - | 4 - 5 | 451 | 4897624 | 150 | |
| BC047774.1 | - | 4 - 5 | 530 | 5761696 | 150 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001429.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10018133.hg.1 | ENST00000372538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018133.hg.1 | ENST00000460899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018133.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018133.hg.1 | TC10001429.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000491270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000491270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000491270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000491270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000491270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018130.hg.1 | ENST00000372538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018130.hg.1 | ENST00000460899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018130.hg.1 | ENST00000536650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018130.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018130.hg.1 | TC10001429.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000491270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000491270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000491270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000491270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000491270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011077.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000372538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000460899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000469299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000470947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000490521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | ENST00000494596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | NM_144589.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175563.hg.1 | TC1000011077.hg.1 | Clariom D TC |

COMTD1
Genomic Map