Assay Details
| Gene Symbol: IDI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004508.3 | NP_004499.2 | 1 - 2 | 499 | 99 | |
| GenBank mRNA | AK303669.1 | - | 1 - 2 | 253 | 99 | |
| BE891119.1 | - | 1 - 2 | 303 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000967.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10012142.hg.1 | ENST00000381344.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012142.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012142.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012142.hg.1 | TC10000967.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012144.hg.1 | ENST00000381344.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012144.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012144.hg.1 | TC10000967.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012150.hg.1 | ENST00000381344.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012150.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012150.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012150.hg.1 | TC10000967.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000427898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000427898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000427898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000427898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000427898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012141.hg.1 | ENST00000381344.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012141.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012141.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012141.hg.1 | TC10000967.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012149.hg.1 | ENST00000381344.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012149.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012149.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012149.hg.1 | TC10000967.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000427898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000427898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000427898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000427898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000427898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000429642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009536.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165162.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165162.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165162.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165162.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165162.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165162.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165162.hg.1 | TC1000009536.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000165161.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165161.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165161.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165161.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165161.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165161.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165161.hg.1 | TC1000009536.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000165160.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165160.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165160.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165160.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165160.hg.1 | ENST00000381344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165160.hg.1 | ENST00000482091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165160.hg.1 | ENST00000491735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165160.hg.1 | NM_004508.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165160.hg.1 | TC1000009536.hg.1 | Clariom D TC |

IDI1
Genomic Map