Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019440.hg.1 |
TC17002917.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019440.hg.1 |
ENST00000251643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019440.hg.1 |
NM_000223.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012399.hg.1 |
ENST00000251643.4 |
Ensembl |