Assay Details
Gene Symbol: AMBN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016519.5 | NP_057603.1 | 5 - 6 | 398 | 90 | |
GenBank mRNA | AF209780.1 | - | 5 - 6 | 380 | 90 | |
AF219994.2 | - | 5 - 6 | 362 | 90 | ||
AF263464.1 | - | 5 - 6 | 372 | 90 | ||
AJ537437.1 | - | 5 - 6 | 372 | 90 | ||
BC106931.1 | - | 5 - 6 | 333 | 40033703 | 90 | |
BC106932.1 | - | 5 - 6 | 333 | 40033705 | 90 | |
BP248666.1 | - | 5 - 6 | 398 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000388.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005289.hg.1 | TC04000388.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005289.hg.1 | ENST00000322937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005289.hg.1 | ENST00000538728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005289.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005290.hg.1 | TC04000388.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005290.hg.1 | ENST00000322937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005290.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005290.hg.1 | ENST00000538728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005290.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153170.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153170.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153170.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153170.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153170.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153170.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153170.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153170.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153170.hg.1 | TC0400007784.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400153169.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153169.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153169.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153169.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153169.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153169.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153169.hg.1 | TC0400007784.hg.1 | Clariom D TC |
