Assay Details
Gene Symbol: AMBN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016519.5 | NP_057603.1 | 9 - 10 | 748 | 77 | |
GenBank mRNA | AF209780.1 | - | 9 - 10 | 730 | 77 | |
AF219994.2 | - | 9 - 10 | 712 | 77 | ||
AF263464.1 | - | 9 - 10 | 77 | |||
AJ537437.1 | - | 9 - 10 | 722 | 77 | ||
AK314918.1 | - | 9 - 10 | 683 | 77 | ||
BC106931.1 | - | 9 - 10 | 683 | 40033703 | 77 | |
BC106932.1 | - | 9 - 10 | 680 | 40033705 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000388.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04005297.hg.1 | TC04000388.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005297.hg.1 | ENST00000322937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005297.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005297.hg.1 | ENST00000538728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005297.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005296.hg.1 | TC04000388.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005296.hg.1 | ENST00000322937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005296.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005296.hg.1 | ENST00000538728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005296.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153176.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153176.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153176.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153176.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153176.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153176.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153176.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153176.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153176.hg.1 | TC0400007784.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400153177.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153177.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153177.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153177.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153177.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153177.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153177.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400153177.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400153177.hg.1 | TC0400007784.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000322937.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000449493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | ENST00000613447.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007784.hg.1 | NM_016519.5 | Refseq |
