Assay Details
Gene Symbol: OR6W1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002140.1 | 1 | 1395 | 98 | ||
GenBank mRNA | BC069382.1 | - | 1 | 986 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | olfactory receptor family 6 subfamily V member 1 | OR6V1 | GPR138 | -9.0kb | Anti-sense | Chr. 7 - 143052341 - 143053282 on NCBI Build GRCh38 |
Downstream | prolactin induced protein | PIP |
GCDFP-15, GCDFP15, GPIP4 |
68.3kb | Anti-sense | Chr. 7 - 143132081 - 143139741 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
