Assay Details
Gene Symbol: FRMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306193.1 | NP_001293122.1 | 10 - 11 | 1103 | 68 | |
NM_194277.2 | NP_919253.1 | 10 - 11 | 1150 | 68 | ||
XM_017029947.1 | XP_016885436.1 | 10 - 11 | 1329 | 68 | ||
XM_017029948.1 | XP_016885437.1 | 7 - 8 | 791 | 68 | ||
XM_017029949.1 | XP_016885438.1 | 6 - 7 | 1112 | 68 | ||
GenBank mRNA | AK125336.1 | - | 10 - 11 | 1150 | 68 | |
AL161984.1 | - | 6 - 7 | 812 | 68 | ||
BC114371.1 | - | 10 - 11 | 1117 | 40079764 | 68 | |
FJ717411.1 | - | 10 - 11 | 927 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X001368.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X018358.hg.1 | TC0X001368.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X018358.hg.1 | ENST00000298542.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X018358.hg.1 | ENST00000370879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X018358.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X018358.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00173553.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00173553.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00173553.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00173553.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00173553.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00173553.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00173553.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00173553.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00173553.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00173553.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00173553.hg.1 | TC0X00010833.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000298542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000370879.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | ENST00000464296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_001306193.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010833.hg.1 | NM_194277.2 | Refseq |
