Assay Details
| Gene Symbol: IARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018060.3 | NP_060530.3 | 4 - 5 | 802 | 93 | |
| GenBank mRNA | AK001188.1 | - | 2 - 3 | 277 | 93 | |
| AK291432.1 | - | 4 - 5 | 611 | 93 | ||
| AY267462.1 | - | 4 - 5 | 813 | 93 | ||
| AY952883.1 | - | 4 - 5 | 95 | |||
| BC040376.1 | - | 4 - 5 | 535 | 93 | ||
| BC047880.1 | - | 3 - 4 | 329 | 93 | ||
| BC137438.1 | - | 4 - 5 | 772 | 9021436 | 93 | |
| D28500.1 | - | 4 - 5 | 643 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001814.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028230.hg.1 | TC01001814.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028230.hg.1 | ENST00000302637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028230.hg.1 | ENST00000366922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028230.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185735.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185735.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185735.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185735.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185735.hg.1 | TC0100011648.hg.1 | Clariom D TC |

IARS2
Genomic Map