Assay Details
| Gene Symbol: IARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018060.3 | NP_060530.3 | 7 - 8 | 1046 | 85 | |
| GenBank mRNA | AK001188.1 | - | 5 - 6 | 521 | 85 | |
| AY267462.1 | - | 7 - 8 | 1057 | 85 | ||
| BC010218.1 | - | 3 - 4 | 226 | 85 | ||
| BC040376.1 | - | 7 - 8 | 779 | 85 | ||
| BC047880.1 | - | 6 - 7 | 573 | 85 | ||
| BC137438.1 | - | 7 - 8 | 1016 | 9021436 | 85 | |
| D28500.1 | - | 7 - 8 | 887 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001814.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01028233.hg.1 | TC01001814.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028233.hg.1 | ENST00000302637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028233.hg.1 | ENST00000366922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028233.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185738.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185738.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185738.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185738.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185738.hg.1 | TC0100011648.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |

IARS2
Genomic Map