Assay Details
| Gene Symbol: IARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018060.3 | NP_060530.3 | 8 - 9 | 1173 | 152 | |
| GenBank mRNA | AK001188.1 | - | 6 - 7 | 648 | 152 | |
| AK001726.1 | - | 2 - 3 | 162 | 152 | ||
| AK291432.1 | - | 8 - 9 | 982 | 152 | ||
| AY267462.1 | - | 8 - 9 | 1184 | 152 | ||
| BC010218.1 | - | 4 - 5 | 353 | 152 | ||
| BC040376.1 | - | 8 - 9 | 906 | 152 | ||
| BC047880.1 | - | 7 - 8 | 700 | 152 | ||
| BC137438.1 | - | 8 - 9 | 1143 | 9021436 | 152 | |
| D28500.1 | - | 8 - 9 | 1014 | 152 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001814.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028234.hg.1 | TC01001814.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028234.hg.1 | ENST00000302637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028234.hg.1 | ENST00000366922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028234.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185739.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185739.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185739.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185739.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185739.hg.1 | TC0100011648.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000366922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000467924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000488777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | ENST00000490891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011648.hg.1 | NM_018060.3 | Refseq |

IARS2
Genomic Map